missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFI Monoclonal Antibody (OTI6F9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806011S
This item is not returnable.
View return policy
Description
CFI Monoclonal antibody specifically detects CFI in Human samples. It is validated for Western BlotSpecifications
| CFI | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P05156 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3426 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI6F9 | |
| Unconjugated | |
| CFI | |
| AHUS3; ARMD13; C3B/C4B inactivator; C3BINA; C3b-INA; C3b-inactivator; Cfi; complement component factor i; complement component I; complement control protein factor I; complement factor I; Complement factor I heavy chain; Complement factor I light chain; factor i; FI; IF; KAF; Konglutinogen-activating factor; light chain of factor I | |
| Human recombinant protein fragment corresponding to amino acids 340-583 of human CFI produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction