Learn More
CFLAR Mouse anti-Human, Clone: OTI2H3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF804485
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
FLIP (CFLAR) is an apoptosis regulator protein which functions as a crucial link between cell survival and cell death pathways in mammalian cells. It acts as an inhibitor of TNFRSF6 mediated apoptosis. A proteolytic fragment (p43) is likely retained in the death-inducing signaling complex (DISC) thereby blocking further recruitment and processing of caspase-8 at the complex. Full length and shorter isoforms have been shown either to induce apoptosis or to reduce TNFRSF-triggered apoptosis. Lacks enzymatic (caspase) activity.Specifications
| CFLAR | |
| Monoclonal | |
| Unconjugated | |
| CFLAR | |
| CASH, CASP8AP1, CLARP, Casper, FLAME, FLAME-1, FLAME1, FLIP, I-FLICE, MRIT, c-FLIP, c-FLIPL, c-FLIPR, c-FLIPS | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8837 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI2H3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O15519 | |
| CFLAR | |
| Human recombit protein fragment corresponding to amino acids 1-272 of human CFLAR produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.