missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CG 428

Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
CG 428 is a potent tropomyosin receptor kinase (TRK) Degrader (uSMITETM) (DC50 = 0.36 nM). CG 428 comprises an analog of the pan-TRK inhibitor GNF-8625 joined by a linker to the cereblon E3 ligase ligand pomalidomide (Cat. No. 6302). CG 428 shows selectivity for TRKA over TRKC and TRKB (Kd values = 1nM, 4.2 nM and 28 nM, respectively). Inhibits growth of KM12 colon cancer cells (IC50 =2.9 nM) and is bioavailable.
Negative control also available, CG 428-Neg (Cat. No. 7426).
Specifications
Specifications
| Chemical Name or Material | 2-(2,6-Dioxo-3-piperidinyl)-4-[[2-[3-[4-[6-[6-[(2 R)-2-(3-fluorophenyl)-1-pyrrolidinyl]imidazo[1,2-b]pyridazin-3-yl]-2-pyridinyl]-1-piperazinyl]-3-oxopropoxy]ethyl]amino]-1 H-isoindole-1,3(2 H)-dione |
| CAS | 2412055-93-5 |
| Quantity | 5 mg |
| Target | Active Degraders |
| Molecular Formula | C43H43FN10O6 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction