missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGB Monoclonal Antibody (OTI12H5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805383S
This item is not returnable.
View return policy
Description
CGB Monoclonal antibody specifically detects CGB in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CGB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WVJ9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 117581 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI12H5 | |
| Unconjugated | |
| TWIST2 | |
| CGB; CGB3; CGB5; CGB7; CGB8; CG-beta; Choriogonadotropin subunit beta; choriogonadotropin subunit beta 3; chorionic gonadotrophin chain beta; chorionic gonadotropin beta 3 subunit; chorionic gonadotropin beta chain; chorionic gonadotropin beta subunit; chorionic gonadotropin beta subunit 3; chorionic gonadotropin chain beta; chorionic gonadotropin human; chorionic gonadotropin, beta polypeptide; FSH-alpha; hCGB; human chorionic gonadotropin; LSH-alpha; TSH-alpha | |
| Full length human recombinant protein of human TWIST2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction