missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGB Monoclonal Antibody (OTI12H5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805383
This item is not returnable.
View return policy
Description
Human chorionic gonadotropin (hCG) is a placental hormone that stimulates secretion of the pregcy-sustaining steroid progesterone. hCG is a member of a family of glycoprotein hormones including pituitary hormones lutropin (luteinizing hormone;LH), follitropin (follicle-stimulating hormone; FSH) and thyrotropin (thyroid-stimulating hormone; TSH). Members of this family are all composed of disulfide-rich heterodimers with two dissimilar subunits namely alpha- and beta-subunits. The alpha subunit is common to all the members of the family within a given species. The beta subunits are distinct in each of the horomones and confer the receptor and biological specificity.Specifications
| CGB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P0DN86 | |
| CGB3 | |
| Human recombit protein fragment corresponding to amino acids 21-165 of human CGB produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI12H5 | |
| Unconjugated | |
| CGB3 | |
| CGB, CGB5, CGB7, CGB8, hCGB | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1082 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction