missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGGBP1 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22325AF488
This item is not returnable.
View return policy
Description
CGGBP1 Polyclonal antibody specifically detects CGGBP1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| CGGBP1 | |
| Polyclonal | |
| Western Blot | |
| 20 kDa CGG-binding protein, CGG triplet repeat binding protein 1, CGG triplet repeat-binding protein 1, CGG-binding protein 1, CGGBPp20-CGGBP, p20-CGG binding protein, p20-CGGBP DNA-binding protein | |
| The immunogen this antibody was made to, maps to a region between residue 100 to 150 of human CGG Triplet Repeat Binding Protein 1 using the numbering given in entry NP_001008391.1 (GeneID 8545). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 8545 | |
| Store at 4°C in the dark. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto