missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CGI-143 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552375
This item is not returnable.
View return policy
Description
Immunogen sequence: SPEVLELRNE SGGHAVPPGS ETHFRVAVVS SRFEGLSPLQ RHRLVHAALA EELGGPVHAL AIQARTPAQW RENSQLDTSP PCLGGNKKTL GT Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
CGI-143 encodes proteins that seem to be involved in cell proliferation or cell-cycle regulation, but the molecular function is still unknown.
Specifications
| CGI-143 | |
| Polyclonal | |
| Unconjugated | |
| BOLA1 | |
| 1810037G04Rik; bolA family member 1; bolA homolog 1; Bola1; bolA-like 1; bolA-like 1 (E. coli); bolA-like protein 1; CGI-143; hBolA; hypothetical protein LOC509530; RGD1310586; zgc:101119 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51027 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y3E2 | |
| BOLA1 | |
| Recombinant protein corresponding to Human CGI-143. Recombinant protein control fragment (Product #RP-89409). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction