missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGI 62 Antibody (OTI4C1), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71896PEATT594
This item is not returnable.
View return policy
Description
CGI 62 Monoclonal specifically detects CGI 62 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| CGI 62 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| ZC2HC1A | |
| Full length recombinant protein of human CGI 62 (NP_057094) produced in E.coli. | |
| 0.1 mL | |
| 51101.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI4C1 | |
| Flow Cytometry | |
| C8orf70, CGI-62, chromosome 8 open reading frame 70, family with sequence similarity 164, member A, hypothetical protein LOC51101 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction