missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CGI58 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542181
This item is not returnable.
View return policy
Description
Peptide sequence: SVIFGARSCI DGNSGTSIQS LRPHSYVKTI AILGAGHYVY ADQPEEFNQK Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 85%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Sheep: 93%.
The protein encoded by this gene belongs to a large family of proteins defined by an alpha/beta hydrolase fold, and contains three sequence motifs that correspond to a catalytic triad found in the esterase/lipase/thioesterase subfamily. It differs from other members of this subfamily in that its putative catalytic triad contains an asparagine instead of the serine residue. Mutations in this gene have been associated with Chanarin-Dorfman syndrome, a triglyceride storage disease with impaired long-chain fatty acid oxidation.
Specifications
| CGI58 | |
| Polyclonal | |
| Unconjugated | |
| ABHD5 | |
| 1300003D03Rik; 1-acylglycerol-3-phosphate O-acyltransferase ABHD5; 2010002J10Rik; ABHD5; abhydrolase domain containing 5; abhydrolase domain-containing protein 5; aCGI58; CDS; CGI 58; CGI58; CGI-58; CGI58 protein; IECN2; IECN5; lipid droplet-binding protein CGI-58; MGC8731; NCIE2; Protein CGI-58; truncated abhydrolase domain-containing protein 5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51099 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8WTS1 | |
| ABHD5 | |
| Synthetic peptide directed towards the C-terminal of human ABHD5 (aa 291-340). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction