Learn More
CGP52432, MedChemExpress
Product Code. 16494886
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16494886
Brand: Medchem Express HY103531
MedChemExpress CGP52432 is a GABAB receptor antagonist, with an IC50 of 85 nM.
CGP52432 is a GABAB receptor antagonist, with an IC50 of 85 nM.
Specifications
Chemical Name or Material CGP52432
Physical Form Solid
CAS 139667-74-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C15H24Cl2NO4P
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 5 mg/mL (13.01 mM; Need ultrasonic and warming) ∣H2O : 4 mg/mL (10.41 mM; ultrasonic and warming and heat to 60°C)
SMILES O=P(C(OCC)OCC)(CCCNCC1=CC=C(Cl)C(Cl)=C1)O
Molecular Weight (g/mol) 384.24
Formula Weight 384.24
Percent Purity 98.17%
Grade Research
Show More Show Less
Research purposes only