missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGREF1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97367PCP
This item is not returnable.
View return policy
Description
CGREF1 Polyclonal antibody specifically detects CGREF1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CGREF1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| cell growth regulator with EF hand domain protein 1, cell growth regulator with EF-hand domain 1, Cell growth regulatory gene 11 protein, CGR11cell growth regulator with EF hand domain 1, hydrophobestin | |
| Produced in rabbits immunized with E. coli-derived Human CGREF1 fragment. | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication | |
| 10669 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction