missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CH25H Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-20218DL594
This item is not returnable.
View return policy
Description
CH25H Polyclonal antibody specifically detects CH25H in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CH25H | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| C25H, cholesterol 25-hydroxylase, Cholesterol 25-monooxygenase, EC 1.14.99.38, h25OH | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CH25H | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 9023 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction