missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ChABC Antibody (1E10), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-96141IR
This item is not returnable.
View return policy
Description
ChABC Monoclonal specifically detects ChABC in Rat, Non-species specific samples. It is validated for Western Blot, ELISA.
Specifications
| ChABC | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| 10000000000 | |
| Western Blot, ELISA | |
| Chondroitinase ABC | |
| Chondroitinase ABC | |
| 0.1 mL | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction