missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ChABC Antibody (6A12), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-96142AF750
This item is not returnable.
View return policy
Description
ChABC Monoclonal specifically detects ChABC in Non-species specific samples. It is validated for Western Blot, ELISA.
Specifications
| ChABC | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| 0.1 mL | |
| Non-species specific | |
| IgG |
| Western Blot, ELISA | |
| 6A12 | |
| Western Blot, ELISA | |
| Chondroitinase ABC | |
| Chondroitinase ABC | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction