missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHAC1 Antibody (OTI1E2), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72191AF488
This item is not returnable.
View return policy
Description
CHAC1 Monoclonal specifically detects CHAC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| CHAC1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CHAC1 | |
| Full length recombinant protein of human CHAC1 (NP_077016) produced in HEK293T cells. | |
| 0.1 mL | |
| 79094.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1E2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| cation transport regulator-like protein 1, ChaC, cation transport regulator homolog 1 (E. coli) | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction