missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHAC1 Monoclonal Antibody (OTI4A11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526310
This item is not returnable.
View return policy
Description
CHAC1 Monoclonal Antibody for Western Blot, ICC/IF
Pro-apoptotic component of the unfolded protein response pathway. May mediate the pro-apoptotic effects of the ATF4-ATF3-DDIT3/CHOP cascade.
Specifications
| CHAC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BUX1 | |
| CHAC1 | |
| Full length human recombinant protein of CHAC1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| OTI4A11 | |
| Unconjugated | |
| CHAC1 | |
| 1810008K03Rik; Blocks Notch protein; botch; cation transport regulator homolog 1; cation transport regulator-like protein 1; ChaC glutathione specific gamma-glutamylcyclotransferase 1; ChaC glutathione-specific gamma-glutamylcyclotransferase 1; ChaC, cation transport regulator 1; ChaC, cation transport regulator homolog 1; ChaC, cation transport regulator-like 1; Chac1; Gamma-GCG 1; gamma-GCT acting on glutathione homolog 1; glutathione-specific gamma-glutamylcyclotransferase 1; Neuroprotective protein 7; Npg7; RGD1307153 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79094 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction