missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Chamber Modification (for Reinforced Shelves or Grids) for UN450
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN450 Oven
Unit Size:
Each
Description
- Includes replacement of standard grids by reinforced grids
Specifications
Specifications
| For Use With (Equipment) | UN450 Oven |
| Type | Chamber Modification |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction