missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Chaperone Protein hchA Protein
Shop All Bio Techne Products
missing translation for 'orderingAttributeHoverText'
Quantity:
0.5 mg
100μg
missing translation for 'unitSize'
0.5mg
100µg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-283 of E.coli Chaperone protein hchA The Recombinant E. coli Chaperone protein hchA Protein is derived from E. coli. The Recombinant E. coli Chaperone protein hchA Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 1.0 mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. 20 mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 0.1 M NaCl, 1 mM DTT |
| Molecular Weight (g/mol) | 33.3 kDa |
| Purification Method | SDS-PAGE |
| Quantity | 0.5 mg |
| Source | Bacteria |
| Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Common Name | Chaperone protein hchA |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction