missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHCHD1 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06218PE
This item is not returnable.
View return policy
Description
CHCHD1 Polyclonal antibody specifically detects CHCHD1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CHCHD1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| C10orf34, C2360, chromosome 10 open reading frame 34, coiled-coil-helix-coiled-coil-helix domain containing 1, coiled-coil-helix-coiled-coil-helix domain-containing protein 1, FLJ25854, Nuclear protein C2360 | |
| Produced in rabbits immunized with E. coli-derived Human CHCHD1 fragment. | |
| 0.1 mL | |
| Cell Biology | |
| 118487 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction