missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHCHD2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17177453
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17177453 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17177453 Supplier Invitrogen™ Supplier No. PA5109524

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TLGHAITGGF SGGSNAEPAR PDITYQEPQG TQPAQQQQPC LYEIKQFLEC AQNQGDIKLC EGFNEVLKQC R.

Transcription factor. Binds to the oxygen responsive element of COX4I2 and activates its transcription under hypoxia conditions (4% oxygen), as well as normoxia conditions (20% oxygen).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHCHD2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CHCHD2
Gene Accession No. Q9D1L0, Q9Y6H1
Gene Alias 16.7kD protein; AAG10; Aging-associated gene 10 protein; AL033347; C7orf17; CHCHD2; coiled-coil-helix-coiled-coil-helix domain containing 2; coiled-coil-helix-coiled-coil-helix domain-containing protein 2; coiled-coil-helix-coiled-coil-helix domain-containing protein 2, mitochondrial; ethanol induced 6; Etohi6; HCV NS2 trans-regulated protein; mitochondria nuclear retrograde regulator 1; MNRR1; NS2TP; PARK22
Gene Symbols CHCHD2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 75-145 of human CHCHD2 (NP_057223.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14004, 316643, 51142
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.