missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHCHD3 Antibody (OTI7G4), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71975B
This item is not returnable.
View return policy
Description
CHCHD3 Monoclonal specifically detects CHCHD3 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| CHCHD3 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| CHCHD3 | |
| Recombinant protein fragment corresponding to amino acids 2-227 of human CHCHD3 (NP_060282) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI7G4 | |
| Western Blot | |
| coiled-coil-helix-coiled-coil-helix domain containing 3, FLJ20420, mitochondrial | |
| Mouse | |
| Affinity Purified | |
| metabolism | |
| 54927.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction