missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHCHD3 Mouse anti-Human, Clone: OTI7G4, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803455S
This item is not returnable.
View return policy
Description
CHCHD3 Monoclonal antibody specifically detects CHCHD3 in Human samples. It is validated for Western BlotSpecifications
| CHCHD3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NX63 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 54927 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI7G4 | |
| Unconjugated | |
| CHCHD3 | |
| 0610041L09Rik; 1700039J09Rik; AW558177; CHCHD3; coiled-coil-helix-coiled-coil-helix domain containing 3; Coiled-coil-helix-coiled-coil-helix domain-containing protein 3; coiled-coil-helix-coiled-coil-helix domain-containing protein 3, mitochondrial; FLJ20420; Mic19; MICOS complex subunit MIC19; MINOS3; mitochondrial inner membrane organizing system 3; PPP1R22; protein phosphatase 1, regulatory subunit 22 | |
| Human recombinant protein fragment corresponding to amino acids 2-227 of human CHCHD3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction