missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHCHD3 Mouse anti-Human, Clone: OTI7G4, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803455
This item is not returnable.
View return policy
Description
Component of the MICOS complex, a large protein complex of the mitochondrial inner membrane that plays crucial roles in the maintece of crista junctions, inner membrane architecture, and formation of contact sites to the outer membrane. Has also been shown to function as a transcription factor which binds to the BAG1 promoter and represses BAG1 transcription. Plays an important role in the maintece of the MICOS complex stability and the mitochondrial cristae morphology.Specifications
| CHCHD3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NX63 | |
| CHCHD3 | |
| Human recombit protein fragment corresponding to amino acids 2-227 of human CHCHD3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI7G4 | |
| Unconjugated | |
| CHCHD3 | |
| MINOS3, Mic19, PPP1R22 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 54927 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction