missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHCHD4 Antibody (6C9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00131474-M01-100ug
This item is not returnable.
View return policy
Description
CHCHD4 Monoclonal antibody specifically detects CHCHD4 in Human samples. It is validated for Western Blot, ELISA
Specifications
| CHCHD4 | |
| Monoclonal | |
| Unconjugated | |
| AAH33775.1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 131474 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 6C9 | |
| In 1x PBS, pH 7.4 | |
| CHDH4, coiled-coil-helix-coiled-coil-helix domain containing 4, Coiled-coil-helix-coiled-coil-helix domain-containing protein 4, FLJ31709, MIA40mitochondrial intermembrane space import and assembly protein 40, mitochondrial intermembrane space import and assembly 40 homolog, TIMM40, translocase of inner mitochondrial membrane 40 homolog | |
| CHCHD4 (AAH33775.1, 1 a.a. ∽ 142 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSYCRQEGKDRIIFVTKEDHETPSSAELVADDPNDPYEEHGLILPNGNINWNCPCLGGMASGPCGEQFKSAFSCFHYSTEEIKGSDCVDQFRAMQECMQKYPDLYPQEDEDEEEEREKKPAEQAEETAPIEATATKEEEGSS | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction