missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHCHD4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99532-50ul
This item is not returnable.
View return policy
Description
CHCHD4 Polyclonal antibody specifically detects CHCHD4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CHCHD4 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| CHDH4, coiled-coil-helix-coiled-coil-helix domain containing 4, Coiled-coil-helix-coiled-coil-helix domain-containing protein 4, FLJ31709, MIA40mitochondrial intermembrane space import and assembly protein 40, mitochondrial intermembrane space import and assembly 40 homolog, TIMM40, translocase of inner mitochondrial membrane 40 homolog | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse CHCHD4. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 131474 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction