missing translation for 'onlineSavingsMsg'
Learn More

CHCHD4 Mouse anti-Human, Rat, Clone: 3E4C6, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66718-1-IG-20UL

 View more versions of this product

Product Code. 16888855

  • € 145.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CHCHD4, a component of human mitochondria, belongs to a protein family whose members share 6 highly conserved cysteine residues constituting a -CXC-CX(9)C-CX(9)C- motif in the C terminus.
Specifications

Specifications

CHCHD4
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8N4Q1
Chchd4
CHCHD4 Fusion Protein Ag15339
20 μL
Primary
Human, Rat
Antibody
IgG1
Western Blot, Immunohistochemistry (Paraffin)
3E4C6
Unconjugated
CHCHD4
CHCHD4, FLJ31709, MIA40
Mouse
Protein G
RUO
131474, 312559
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.