missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHCHD4 Polyclonal Antibody

Product Code. 15774911 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15774911 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15774911 Supplier Invitrogen™ Supplier No. PA557015

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ELVADDPNDP YEEHGLILPN GNINWNCPCL GGMASGPCGE QFKSAFSCFH YSTEEIKGSD CVDQFRAMQE CMQKYPDL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

CHCHD4, a component of human mitochondria, belongs to a protein family whose members share 6 highly conserved cysteine residues constituting a -CXC-CX(9)C-CX(9)C- motif in the C terminus (Hofmann et al., 2005).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHCHD4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHCHD4
Gene Accession No. Q8N4Q1
Gene Alias 2410012P20Rik; 2810014D17Rik; AI838740; CHCHD4; coiled-coil-helix-coiled-coil-helix domain containing 4; coiled-coil-helix-coiled-coil-helix domain-containing protein 4; MIA40; mitochondrial intermembrane space import and assembly 40 homolog; mitochondrial intermembrane space import and assembly protein 40; TIMM40; translocase of inner mitochondrial membrane 40 homolog
Gene Symbols CHCHD4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CHCHD4. Recombinant protein control fragment (Product #RP-96401).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131474
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.