missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHCHD5 Antibody (OTI3H3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-03608
This item is not returnable.
View return policy
Description
CHCHD5 Monoclonal specifically detects CHCHD5 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CHCHD5 | |
| Monoclonal | |
| 0.35 mg/mL | |
| Western Blot 1:200, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150 | |
| CHCHD5 | |
| Human recombinant protein fragment corresponding to amino acids 1-110 of human CHCHD5(NP_115685) produced in E.coli. | |
| Affinity Purified | |
| RUO | |
| 84269 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI3H3 | |
| Unconjugated | |
| C2orf9, chromosome 2 open reading frame 9, coiled-coil-helix-coiled-coil-helix domain containing 5, coiled-coil-helix-coiled-coil-helix domain-containing protein 5, FLJ39671, MGC11104 | |
| Mouse | |
| 12.2 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction