missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHCHD5 Monoclonal Antibody (OTI6G6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502373S
This item is not returnable.
View return policy
Description
CHCHD5 Monoclonal antibody specifically detects CHCHD5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CHCHD5 | |
| Monoclonal | |
| 0.6 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BSY4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84269 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6G6 | |
| Unconjugated | |
| CHCHD5 | |
| 1110027L01Rik; AW045710; C2orf9; Chchd5; coiled-coil-helix-coiled-coil-helix domain containing 5; coiled-coil-helix-coiled-coil-helix domain-containing protein 5; MIC14; mitochondrial intermembrane space cysteine motif protein of 14 kDa homolog | |
| Human recombinant protein fragment corresponding to amino acids 1-110 of human CHCHD5 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction