missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576896
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The CHD family of proteins is characterized by the presence of chromo domains and SNF2-related helicase/ATPase domains. CHD genes alter gene expression possibly by modification of chromatin structure thus altering access of the transcriptional apparatus to its chromosomal DNA template. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
| CHD2 | |
| Polyclonal | |
| Unconjugated | |
| CHD2 | |
| 2810013C04Rik; 2810040A01Rik; 5630401D06Rik; AI851092; ATP-dependent helicase CHD2; BC029703; chd2; CHD-2; chromodomain helicase DNA binding protein 2; chromodomain-helicase-DNA-binding protein 2; DKFZp547I1315; DKFZp686E01200; DKFZp781D1727; Down syndrome cell adhesion molecule; EEOC; FLJ38614; LOW QUALITY PROTEIN: chromodomain-helicase-DNA-binding protein 2; si:ch211-196l7.5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1106, 244059, 308738 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| E9PZM4, O14647 | |
| CHD2 | |
| Recombinant full length Human CHD2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction