missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD4 Antibody (CHD4 3F2/4), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50163APC
This item is not returnable.
View return policy
Description
CHD4 Monoclonal specifically detects CHD4 in Human, Mouse samples. It is validated for Flow Cytometry, Knockout Validated.
Specifications
| CHD4 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| CHD4 | |
| Synthetic peptide (N- and C- terminal peptides were used for immunisation. 1. ASGLGSPSPCSAGSEEEDM & 2. CSRLANRAPEPPPQQVAQQQ. Which peptide the antibody recognises has not been tested) | |
| 0.1 mL | |
| Immune System Diseases, Immunology | |
| 1108 | |
| Store at 4C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| CHD4 3F2/4 | |
| Flow Cytometry | |
| ATP-dependent helicase CHD4, CHD-4, chromodomain helicase DNA binding protein 4, chromodomain-helicase-DNA-binding protein 4, DKFZp686E06161, EC 3.6.1, EC 3.6.4.12, Mi-2 autoantigen 218 kDa protein, Mi-2b, Mi2-BETA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction