missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD4 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33432-100ul
This item is not returnable.
View return policy
Description
CHD4 Monoclonal antibody specifically detects CHD4 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| CHD4 | |
| Monoclonal | |
| ELISA Recommended starting concentration is 1 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| ATP-dependent helicase CHD4, CHD-4, chromodomain helicase DNA binding protein 4, chromodomain-helicase-DNA-binding protein 4, DKFZp686E06161, EC 3.6.1, EC 3.6.4.12, Mi-2 autoantigen 218 kDa protein, Mi-2b, Mi2-BETA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1520-1700 of human CHD4 (NP_001264.2).,, Sequence:, ELAEVEENKKMSQPGSPSPKTPTPSTPGDTQPNTPAPVPPAEDGIKIEENSLKEEESIEGEKEVKSTAPETAIECTQAPAPASEDEKVVVEPPEGEEKVEKAEVKERTEEPMETEPKGAADVEKVEEKSAIDLTPIVVEDKEEKKEEEEKKEVMLQNGETPKDLNDEKQKKNIKQRFMFNI | |
| 100 μL | |
| Immune System Diseases, Immunology | |
| 1108 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction