missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD4 Antibody (SR1411), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21737-50ul
This item is not returnable.
View return policy
Description
CHD4 Monoclonal antibody specifically detects CHD4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
Specifications
| CHD4 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| SR1411 | |
| Western Blot 1:1000-1:5000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| ATP-dependent helicase CHD4, CHD-4, chromodomain helicase DNA binding protein 4, chromodomain-helicase-DNA-binding protein 4, DKFZp686E06161, EC 3.6.1, EC 3.6.4.12, Mi-2 autoantigen 218 kDa protein, Mi-2b, Mi2-BETA | |
| A synthesized peptide derived from human CHD4 (Uniprot #: Q14839) | |
| 50 μg | |
| Immune System Diseases, Immunology | |
| 1108 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction