missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHD7 Monoclonal Antibody (772503)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561807
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Chromodomain helicase DNA binding protein 7 (CHD7) is a gene located on chromosome 8q12.2 that encodes a member of the chromodomain helicase DNA-binding (CHD) family, which plays a pivotal role in chromatin remodeling and gene expression regulation. CHD7 contains multiple domains that facilitate its involvement in modifying chromatin structure, thereby influencing the accessibility of DNA for transcription factors and other regulatory proteins. The protein is integral to various developmental processes, including neural crest formation, thus affecting tissues such as the nervous system, skeletal structures, and sensory organs. Mutations in CHD7 are associated with CHARGE syndrome, a multisystem disorder characterized by congenital anomalies including coloboma, heart defects, and ear abnormalities. CHD7's role in chromatin dynamics makes it crucial for normal embryonic development and cellular differentiation. Ongoing research is focused on uncovering CHD7's broader implications in developmental biology and its potential as a therapeutic target in chromatin-related disorders.
Specifications
| CHD7 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9P2D1 | |
| CHD7 | |
| E. coli-derived recombinant human CHD7, Ala263-Gln457. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry | |
| 772503 | |
| Unconjugated | |
| CHD7 | |
| A730019I05Rik; ATP-dependent helicase CHD7; CHARGE association; CHD7; CHD-7; chromodomain helicase DNA binding protein 7; chromodomain helicase DNA binding protein 7 isoform CRA_e; chromodomain-helicase-DNA-binding protein 7; CRG; Cycn; Cyn; Dz; Edy; Flo; GENA 47; Gena 52; GENA 60; HH5; IS3; KAL5; KIAA1416; Lda; metis; Mt; Obt; Todo; WBE1; Whi | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 55636 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction