Learn More
Cole-Parmer™ Check Valves with Pipe Thread Connections
Click to view available options
Maximum Pressure:
100 psi
125 psi
150 psi
Unit Size:
Each
Description
- Check valve
- With pipe thread connections
- Available with 1/4 or 1/2 in. NPT(F) Connections
Specifications
Specifications
| Material | Polypropylene, Viton |
| Inlet | 1/2 in. NPT(F) |
| Outlet Port | 1/2 in. NPT(F) |
| Maximum Pressure | 100 psi |
| Max. Operating Temperature (English) | 180°F |
| Max. Operating Temperature (Metric) | 81°C |
| Product Type | Check Valve |