Learn More
Hanna Instruments™ Checktemp™ Thermometer
Product Code. 10083123
Click to view available options
Temperature Range (Metric):
-50°C to +220°C
Unit Size:
Each
Product Code. 10083123
Brand: Hanna Instruments™ HI15100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Product Type Thermometer
Max. Temperature (Metric) 220°C
Min. Temperature (Metric) -50°C
Temperature Range (Metric) -50°C to +220°C
Accuracy ±0.3°C