Learn More
Masterflex™ Chem-Durance™ Bio I/P™ High-Performance Precision Pump Tubing
Enhanced pumping life and durability—especially under pressure
€ 1345.00 - € 1410.00
Specifications
Autoclavable | Yes |
---|---|
Material | Chem-Durance Bio |
Min. Operating Temperature (Metric) | -60°C |
Color | Opaque |
MaxOperatingTempMetric | 74°C |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
11876953
|
Masterflex™
06442-88 |
88 |
€ 1345.00
Each |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
11886953
|
Masterflex™
06442-89 |
89 |
€ 1410.00
Each |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
Description
High Performance Tubing is the best choice for applications involving pressure, suction lift, viscous fluids or long tubing life. Thicker wall compared to precision pump tubing. Performs in a wide range of chemical applications limiting the number of tubing formulations you need to keep on hand. Low spallation maintains purity of fluid. Flexible outer jacket acts as an insulating layer to absorb and dissipate heat from the rollers and extending the life of the tubing.- Plasticizer-free, smooth bore liner is hydrophobic and resists the absorption/adsorption of aqueous fluids Complies with USP Class VI, FDA, REACH, RoHS
- Excellent chemical compatibility and biocompatibility
- Excellent pump life and durability under pressure
- Excellent chemical resistance and long pumping life
- Meets USP Class VI requirements
Specifications
Yes | |
-60°C | |
74°C |
Chem-Durance Bio | |
Opaque |