Learn More
Welch™ Chemical Resistant Diaphragm Pump
Product Code. 12602905 Shop All Welch Ilmvac Products
Change view
Click to view available options
Model:
MPC 1801 Z
MPC 2401 E
MPC 601 T - EF
Dimensions (L x W x H):
260 x 390 x 170 mm
540 x 300 x 240 mm
Flow Rate:
13.3 m³/hr.
17 m³/hr.
4.9 m³/hr.
No. of Pump Heads:
4
8
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Dimensions (L x W x H) Flow Rate No. of Pump Heads unitSize
12602905 MPC 1801 Z 540 x 300 x 240 mm 13.3 m³/hr. 8 Each
11852263 MPC 601 T - EF 260 x 390 x 170 mm 4.9 m³/hr. 4 Each
12692895 MPC 2401 E 540 x 300 x 240 mm 17 m³/hr. 8 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 12602905 Supplier Welch™ Supplier No. 412782
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
The MPC chemical duty diaphragm pumps allow aggressive solvent and acid vapors to be pumped.
  • The MPC chemical duty diaphragm pumps use PTFE and other fluorinated plastics for the wetted parts to allow aggressive solvent and acid vapors to be pumped. If the pump should handle harsh chemical vapors, then the superior chemical resistant X2 version with higher grade seals and thicker coatings are the best choice. The diaphragm series from Welch is well known for their quiet operation and outstanding running smoothness. This is a result of continuous product improvements and close relationships with our customers. With their low weight and compact design, users can easily move pumps to different locations for use on different applications.
  • Depending on the specific requirements, Welch provides 1, 2, 4, and 8-head diaphragm pumps. They are available in one or multistage configuration with controlled (ecoflex) or uncontrolled pumping speed. Ecoflex diaphragm pumps are equipped with a state-of-the-art vacuum control which allows the pumping speed to be varied. The vacuum can be adjusted precisely to individual needs with the result that the process runs more efficiently and is reproducible. The additional pressure reduction allows the distillation of mixtures almost automatically and without fractionated operation. Ecoflex diaphragm pumps guarantee a positive economic and ecological balance with the result that operation costs can be reduced by up to 80%.
  • Applications: Rotary evaporation, Vacuum oven, Industrial Applications, Desiccation, Distillation, Filtration, Gel drying, Concentrator, Gas sampling, Drying, Degassing
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W x H) 540 x 300 x 240 mm
Material PTFE
Model MPC 1801 Z
Noise Level < 44 dB
Weight (English) 72.3 lb.
Weight (Metric) 32.8 kg
Flow Rate 13.3 m³/hr.
For Use With Rotary Evaporation, Vacuum Oven, Industrial Applications, Desiccation, Distillation, Filtration, Gel Drying, Concentrator, Gas Sampling, Drying, Degassing
Frequency 50/60 Hz
Pump Type Chemical Resistant Diaphragm Pump
No. of Pump Heads 8
No. of Stages 2
Maximum Pressure <8 torr
Voltage 230 V