missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Schneider Gerd™ Chemical Shovel
Use these scoops for a variety of applications. Schneider Gerd™ Chemical Shovels are available in cast aluminum and stainless steel.
€ 4.25 - € 27.30
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10403274
|
Schneider Gerd™
527 |
310 mm |
€ 27.30
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
10683585
|
Schneider Gerd™
522 |
185 mm |
€ 9.00
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
10506864
|
Schneider Gerd™
524 |
250 mm |
€ 16.00
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
10602635
|
Schneider Gerd™
526 |
290 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10403004
|
Schneider Gerd™
520 |
145 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10181984
|
Schneider Gerd™
523 |
210 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10673585
|
Schneider Gerd™
521 |
155 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Metal lab scoops
- For transferring or weighing solids
Specifications
Chemical Shovel |