Learn More
3M™ Chemical Spill Kits
Click to view available options
Capacity (Metric):
26 L
Unit Size:
Each
Description
- Ideal for rapid response to chemical spill containment and cleanup
- Cleans up hazardous chemical spills, drips and leaks
- Suitable for use with most chemicals
- Promotes safety on the site
- Easy to dispose of
- A selection of 3M chemical sorbents of varying formats is included as well as cleanup bags
Specifications
Specifications
| Capacity (Metric) | 26 L |
| Type | Chemical Spill Kit |
| For Use With (Application) | Cleaning up hazardous chemical spills, drips, leaks |
| Includes | 10 x P110 sheets, 5 x P300 pillows, 5 x P200 mini-brooms, 2x disposal bags and ties |
| Packaging Quantity | 1 Ea. |
| Thickness | 40 mm |
| Width (Metric) | 400 mm |