missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Chemical Test Kit

Product Code. 10637554
Change view
Click to view available options
For Use With (Application):
Ammonia Determination in Fresh Water
Carbon Dioxide Determination
Iron (Fe2+ and Fe3+) Determination
Phosphate Determination
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
10637554 Carbon Dioxide Determination Each
10504784 Iron (Fe2+ and Fe3+) Determination Each
10480684 Ammonia Determination in Fresh Water Each
10634155 Phosphate Determination Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10637554 Supplier Hanna Instruments™ Supplier No. HI3818
  • ONLINE EXCLUSIVE PRICE
    € 66.73
    € 50.15 / Pack of 1
    Save € 16.58
    24% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Chemical Test Kit features pre-made, pre-measured reagents for fast, accurate determinations with high-resolution readings based on accepted titrations methods.

These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.

  • Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
  • High resolution readings for fast, accurate determinations
  • Replacement reagents available: No need to buy a new kit when reagents are exhausted

Specifications

Analytes Phenolphthalein
For Use With (Application) Carbon Dioxide Determination
For Use With (Equipment) Water samples
Includes 1 (10mL) dropper bottle phenolphthalein indicator, 120mL bottle carbon dioxide reagent, 10mL calibrated vessel, 50mL calibrated vessel and calibrated syringe
No. of Tests 110
Quantity 1 Kit
Range 0.0 to 10.0mg/L, 0.0 to 50.0mg/L, 0 to 100mg/L
Sufficient For Three ranges
Test Method Titration
Type Chemical Test Kit
compliance-icons compliance-icons compliance-icons
Product Identifier
  • CARBON DIOXIDE CTK
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use powder to extinguish.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, phenolphtalein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.