Learn More
Cole-Parmer™ Stuart™ Chemically Resistant ECTFE Bottle Top Adaptor
Chemically resistant ECTFE bottle top adaptor compatible with the Stuart™ range of bottle top dispensers
Brand: Cole-Parmer™ 10116-94
Additional Details : Weight : 0.01000kg
Description
- For use with a variety of Stuart™ bottle top dispensers
- Chemically resistant ECTFE
- Available in 38mm or 40mm thread size
Specifications
Adapter | |
ECTFE | |
Pressmatic Bottle Top Dispenser | |
40 mm |