missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHES1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-04051UV
This item is not returnable.
View return policy
Description
CHES1 Polyclonal antibody specifically detects CHES1 in Human samples. It is validated for Western Blot
Specifications
| CHES1 | |
| Polyclonal | |
| Western Blot | |
| C14orf116, Checkpoint suppressor 1PRO1635, CHES1forkhead box protein N3, chromosome 14 open reading frame 116, forkhead box N3 | |
| The immunogen recognized by this antibody maps to a region between residue 440 and 490 of human Forkhead Box N3 using the numbering given in entry NP_001078940.1 (GeneID 1112). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 1112 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction