missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHGA Mouse anti-Human, Mouse, Clone: OTI2E12, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506098S
This item is not returnable.
View return policy
Description
CHGA Monoclonal antibody specifically detects CHGA in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| CHGA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P10645, P26339 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1113, 12652 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2E12 | |
| Unconjugated | |
| Chga | |
| AL-11; AL26; Beta-granin; betagranin (N-terminal fragment of chromogranin A); catestatin; CgA; CHGA; ChrA; chromofungin; chromogranin A; chromogranin A (parathyroid secretory protein 1); chromogranin A precursor; chromogranin-A; CTNNB; DKFZp686D02253; EA-92; ER-37; ES-43; FLJ25606; FLJ37923; GE-25; GR-44; GV-19; LF-19; LOW QUALITY PROTEIN: chromogranin-A; MGC105435; Pancreastatin; Parastatin; parathyroid secretory protein 1; p-Glu serpinin precursor; pituitary secretory protein I; Serpinin; Serpinin-RRG; SL21; SP-I; SS-18; Vasostatin I; Vasostatin II; Vasostatin-1; Vasostatin-2; WA-8; WE-14 | |
| Full length human recombinant protein of human CHGA produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction