missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHIC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111741
This item is not returnable.
View return policy
Description
Immunogen sequence: SIEKLLEWEN NRLYHKLCLH WRLSKRKCET N Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
This gene encodes a member of the CHIC family of proteins. The encoded protein contains a cysteine-rich hydrophobic (CHIC) motif, and is localized to vesicular structures and the plasma membrane. This gene is associated with some cases of acute myeloid leukemia.
Specifications
| CHIC2 | |
| Polyclonal | |
| Unconjugated | |
| CHIC2 | |
| 1700081B18Rik; 4930502K01Rik; BrX-like translocated in leukemia; BTL; Chic2; cysteine rich hydrophobic domain 2; cysteine-rich hydrophobic domain 2; cysteine-rich hydrophobic domain 2 protein; cysteine-rich hydrophobic domain-containing protein 2; cystein-rich hydrophobic domain 2; MGC21173; zgc:55670 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26511 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UKJ5 | |
| CHIC2 | |
| Recombinant Protein corresonding to Human CHIC2. Recombinant protein control fragment (Product #RP-108667). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction