missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chicken Anemia Virus VP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117707
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
Self-assembles to form the virion icosahedral capsid with a T=1 symmetry. This very small capsid (25 nm in diameter) allows the virus to be very stable in the environment and resistant to some disinfectants, including detergents. Essential for the initial attachment to host receptors. After attachment, the virus is endocytosed and traffics to the nucleus. The capsid protein binds and transports the viral genome and Rep across the nuclear envelope.
Specifications
| Chicken Anemia Virus VP1 | |
| Polyclonal | |
| Unconjugated | |
| VP1 | |
| Recombinant Chicken anemia virus Capsid protein (288-369aa). | |
| 100 μL | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
| Western Blot | |
| 2.79 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Rabbit | |
| Protein G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction