missing translation for 'onlineSavingsMsg'
Learn More

CNP, Chicken, Polyclonal Antibody, Abnova™

Product Code. 16147020
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16147020 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16147020 Supplier Abnova Supplier No. PAB29066.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chicken polyclonal antibody raised against recombinant Human CNP.

Specifications

Antigen CNP
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against recombinant Human CNP.
Dilution Immunocytochemistry(1:1000-1:2000) Immunohistochemistry(1:1000-1:2000) Western Blot(1:2000-1:5000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.02% sodium azide)
Gene Accession No. NP_149124
Gene Alias CNP1
Gene Symbols CNP
Host Species Chicken
Immunogen A KLH-conjugated synthetic peptide corresponding to a region of CNP gene product shared between the human (NP_149124, NCBI) and mouse (P16330, NCBI) sequences.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1267/12799
Target Species Human, Mouse
Content And Storage Store at 4°C and avoid from light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.