Learn More
KRT20, Chicken, Biotin, Polyclonal Antibody, Abnova™
Chicken polyclonal antibody against synthetic peptide of KRT20
Brand: Abnova PAB27966.50ug
Description
The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. This cytokeratin is a major cellular protein of mature enterocytes and goblet cells and is specifically expressed in the gastric and intestinal mucosa. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21. [provided by RefSeq
Sequence: LTVEADLQGLNKSpecifications
| KRT20 | |
| Polyclonal | |
| Biotin | |
| In Phosphate-Buffered Saline with 0.02% Sodium Azide. | |
| CD20/CK20/K20/KRT21/MGC35423 | |
| Chicken | |
| Antigen affinity purification | |
| RUO | |
| 54474 | |
| Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles. | |
| IgY |
| Immuno-MRM (Multiple Reaction Monitoring) | |
| 1 mg/mL | |
| Chicken polyclonal antibody against synthetic peptide of KRT20 | |
| KRT20 | |
| KRT20 | |
| A Synthetic peptide corresponding to amino acids of KRT20 | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only