missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEFL, Chicken, Polyclonal Antibody, Abnova™
Chicken polyclonal antibody raised against recombinant Human NEFL.
Brand: Abnova PAB29082.300ug
This item is not returnable.
View return policy
Specifications
| NEFL | |
| Polyclonal | |
| Chicken polyclonal antibody raised against recombinant Human NEFL. | |
| In PBS, pH 7.2 (0.2% BSA, 0.02% sodium azide) | |
| NEFL | |
| Three different KLH-conjugated synthetic peptides corresponding to different regions of NEFL gene product, shared between the human (P07196, NCBI) and mouse (P08551, NCBI) protein sequences. | |
| 300 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry | |
| Unconjugated | |
| Immunocytochemistry(1:1000-1:2000) Immunohistochemistry(1:1000-1:2000) The optimal working dilution should be determined by the end user. | |
| CMT1F/CMT2E/FLJ53642/NF-L/NF68/NFL | |
| Chicken | |
| Antigen affinity purification | |
| RUO | |
| 4747/18039 | |
| Store at 4°C and avoid from light. Aliquot to avoid repeated freezing and thawing. |
|
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction