missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSRP1, Chicken, Polyclonal Antibody, Abnova™
Chicken polyclonal antibody raised against synthetic peptide of NSRP1.
Brand: Abnova PAB25732.100ug
This item is not returnable.
View return policy
Specifications
| NSRP1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) Immunofluorescence (20 ug/mL) The optimal working dilution should be determined by the end user. | |
| NSRP1 | |
| NSRP1 | |
| A synthetic peptide corresponding to 16 amino acids at C-terminus of human NSRP1. | |
| 100 μg | |
| Primary | |
| At least three isoforms are known to exist; this antibody will detect all three. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgY |
| ELISA, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Chicken polyclonal antibody raised against synthetic peptide of NSRP1. | |
| In PBS (0.02% sodium azide) | |
| CCDC55/HSPC095/NSrp70 | |
| Chicken | |
| Peptide affinity purification | |
| RUO | |
| 84081 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction